missing translation for 'onlineSavingsMsg'
Learn More

(+)-Biotin N-succinimidyl ester, 98%

Catalog No. p-7059520
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4477101 0.25 g
AA4477103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4477101 Supplier Thermo Scientific Chemicals Supplier No. 04477101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

In coupling reactions to amino acids, peptides and proteins through primary amines | CAS: 35013-72-0 | C14H19N3O5S | 341.382 g/mol

(+)-Biotin N-succinimidyl ester is used in coupling reactions to amino acids, peptides and proteins through primary amines. It is used as amino reactive biotin reagent and in the preparation of biotinylated surfaces or polypeptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35013-72-0
Molecular Formula C14H19N3O5S
Molecular Weight (g/mol) 341.382
MDL Number MFCD00078531
InChI Key YMXHPSHLTSZXKH-RVBZMBCESA-N
Synonym +-biotin n-hydroxysuccinimide ester, biotin-nhs, biotin nhs, 2,5-dioxopyrrolidin-1-yl 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanoate, n-hydroxysuccinimidobiotin, d-biotinyl-osu, biotin-osu, n-succinimidyl d-biotinate, bhse, biotin n-hydroxysuccinimide ester
PubChem CID 6710714
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoate
SMILES C1CC(=O)N(C1=O)OC(=O)CCCCC2C3C(CS2)NC(=O)N3

Specifications

Melting Point 212°C to 215°C
Color White
Quantity 0.25 g
Beilstein 4720781
Solubility Information Soluble in Dimethylformamide ?50mg/mL (Stable for at least one month in dry DMF). Soluble in Dimethyl sulfoxide(DMSO).
Formula Weight 341.4
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (+)-Biotin N-succinimidyl ester
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only