missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (+)-Biotin N-hydroxysuccinimide ester, 98%

CAS: 35013-72-0 | C14H19N3O5S | 341.39 g/mol

Supplier:  Thermo Scientific Chemicals 439302500

 Afficher plus de versions de ce produit

Numéro de catalogue. AC439302500


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

(+)-Biotin N-hydroxysuccinimide ester
97.5
202.3°C to 202.7°C
48.03 to 50.49% C, 12.00 to 12.62% N
98%
C14H19N3O5S
+-biotin n-hydroxysuccinimide ester, biotin-nhs, biotin nhs, 2,5-dioxopyrrolidin-1-yl 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanoate, n-hydroxysuccinimidobiotin, d-biotinyl-osu, biotin-osu, n-succinimidyl d-biotinate, bhse, biotin n-hydroxysuccinimide ester
C1CC(=O)N(C1=O)OC(=O)CCCCC2C3C(CS2)NC(=O)N3
341.39
341.39
Powder
35013-72-0
100.0
White
Authentic
Glass bottle
250 mg
YMXHPSHLTSZXKH-RVBZMBCESA-N
(2,5-dioxopyrrolidin-1-yl) 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoate
6710714
98%

Identifiants chimiques

35013-72-0
341.39
+-biotin n-hydroxysuccinimide ester, biotin-nhs, biotin nhs, 2,5-dioxopyrrolidin-1-yl 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanoate, n-hydroxysuccinimidobiotin, d-biotinyl-osu, biotin-osu, n-succinimidyl d-biotinate, bhse, biotin n-hydroxysuccinimide ester
(2,5-dioxopyrrolidin-1-yl) 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoate
C14H19N3O5S
YMXHPSHLTSZXKH-RVBZMBCESA-N
6710714
C1CC(=O)N(C1=O)OC(=O)CCCCC2C3C(CS2)NC(=O)N3
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:

GHS P Statement:
Do not get in eyes, on skin, or on clothing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

FDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only