Learn More
Innovatek BioStrip™ Urinalysis Reagent Strips
Catalog No. 361041271
Change view
Click to view available options
Model:
BioStrip™ 2P
BioStrip™ U10
BioStrip™ U7
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
361041271 BioStrip™ 2P
361016428 BioStrip™ U7
361016429 BioStrip™ U10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 361041271 Supplier Innovatek Supplier No. 41922P
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Innovatek BioStrip™ Urinalysis Reagent Strips provide consistent, easily interpreted results
  • Excellent quality with unsurpassed economy
  • Popular test configurations
  • Consistent results that are easily interpreted
  • Long shelf life, desiccant keeps strips fresh
Specifications
Detectable Analytes Protein, Glucose
Type Reagent Strip
For Use With (Application) Urinalysis
Includes Desiccant
Model BioStrip™ 2P
Quantity 100 Test Strips/Bottle