Learn More
Innovatek BioStrip™ Urinalysis Reagent Strips
Innovatek BioStrip™ Urinalysis Reagent Strips provide consistent, easily interpreted results
Supplier: Innovatek 4192U7
Description
- Excellent quality with unsurpassed economy
- Popular test configurations
- Consistent results that are easily interpreted
- Long shelf life, desiccant keeps strips fresh
Chemical Identifiers
| Reagent Strip | |
| Urinalysis | |
| Desiccant | |
| 100 Test Strips/Bottle |
Specifications
| Glucose, Ketone, Protein, pH, Blood, Nitrite, Leukocyte | |
| Urinalysis | |
| BioStrip™ U7 |
| Reagent Strip | |
| Desiccant | |
| 100 Test Strips/Bottle |