missing translation for 'onlineSavingsMsg'
Learn More

Innovatek BioStrip™ Pregnancy Test Strips

Catalog No. 361011109
Change view
Click to view available options
Quantity:
25 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361011109 25 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361011109 Supplier Innovatek Supplier No. 42205

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Innovatek BioStrip™ Pregnancy Test Strips provide results within 5 minutes

  • Cost effective and minimal packaging
  • Desiccant in cap keeps strips fresh
  • Dip and read in 5 minutes

Specifications

Format Dipstick
For Use With (Application) Pregnancy Tests
Quantity 25 Tests/Kit
Test Time 5 min.