missing translation for 'onlineSavingsMsg'
Learn More

Innovatek BioStrip™ Ovulation Prediction Test

Catalog No. 361011110
Change view
Click to view available options
Quantity:
6 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361011110 6 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361011110 Supplier Innovatek Supplier No. 4220 13

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Innovatek BioStrip™ Ovulation Prediction Test detects elevated levels of Luteinizing Hormone (LH) in urine

  • Qualitative immunoassay detects elevated levels of Luteinizing Hormone (LH) in urine, indicating periods of optimal fertility
  • Individually foil wrapped dipstick strip format
  • Each kit is for one cycle

Specifications

Detectable Analytes Luteinizing Hormone (LH)
Format Dipstick
For Use With (Application) Ovulation Tracking
Packaging Type Individually Packaged
Quantity 6 Tests/Kit