Learn More
Research Products Corporation Bio-Safe II Complete Counting Cocktail, 4 Liters
Catalog No. M111195
Encompass
Our highest performance biodegradable cocktail for counting a wide range of samples including: aqueous solutions, biological materials, salt-buffer combinations and organic solutions. Bio-Safe II has a sample holding capacity of up to 25% aqueous volume within a clear, stable region. This large capacity can actually reduce the amount of scintillation fluid required which becomes important when using miniature vials. Bio-Safe II will not permeate plastic scintillation vials even over extended storage periods.
Catalog No. M111195 Supplier Research Products Corporation Supplier No. RPI 111195
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Our highest performance biodegradable cocktail for counting a wide range of samples including: aqueous solutions, biological materials, salt-buffer combinations and organic solutions. Bio-Safe II has a sample holding capacity of up to 25% aqueous volume within a clear, stable region. This large capacity can actually reduce the amount of scintillation fluid required which becomes important when using miniature vials. Bio-Safe II will not permeate plastic scintillation vials even over extended storage periods.