Learn More
FEATHER Biopsy Punch
Feather Biopsy Punch offers excellent sharpness by ultra-precision sharpening technology, which originates in surgical blade manufacturing.
Supplier: FEATHER FBP30
Description
- Made in Japan
- Packaged with safety cover
- Anti-roll hexagon design
- Completely-sealed package by special film
- 20/Box
- Sterilized by Gamma radiation
- Single use only
Chemical Identifiers
| Biopsy Punch | |
| Single-use | |
| Sterile |
Specifications
| Biopsy Punch | |
| Single-use | |
| Sterile | |
| 3.0 mm |