Learn More
FEATHER Biopsy Punch
Catalog No. 361046711
Change view
Click to view available options
Diameter (Metric):
1.5 mm
3.0 mm
4.0 mm
6.0 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
361046711 1.5 mm
361046712 3.0 mm
361046713 4.0 mm
361046714 -
361046715 6.0 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 361046711 Supplier FEATHER Supplier No. FBP15
Feather Biopsy Punch offers excellent sharpness by ultra-precision sharpening technology, which originates in surgical blade manufacturing.
  • Made in Japan
  • Packaged with safety cover
  • Anti-roll hexagon design
  • Completely-sealed package by special film
  • 20/Box
  • Sterilized by Gamma radiation
  • Single use only
Specifications
Product Type Biopsy Punch
Disposable Single-use
Sterility Sterile
Diameter (Metric) 1.5 mm