Learn More
StatLab BioMesh Cassettes
Catalog No. STLHS501
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Pink
Red
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
STLHS501 Blue
STLHS500 White
STLHS502 Yellow
STLHS503 Green
STLHS504 Pink
STLHS505 Tan
STLHS507 Gray
STLHS509 Aqua
STLHS510 Red
STLHS511 Lilac
STLHS512 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. STLHS501 Supplier StatLab Supplier No. HS501
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
StatLab BioMesh Cassettes keep small biopsy samples secure and are ideal for automated cassette printers.
  • Lids separate, ideal for small, fragile specimens
  • Excellent for use with Tissue Guard processing gel
  • Keep small biopsies and fragile specimens secure with these fine mesh cassettes that eliminate the need for messy sponges, bags and paper
  • Rounded BioMesh edges ensure no tissue fragments are lost in the corners
Specifications
Quantity 250 Pk.
Color Blue