Learn More
Biomedical Polymers Hemodrop Blood Dispenser
Catalog No. 22544233
Change view
Click to view available options
Quantity:
250 Pack
Catalog No. Quantity
22544233 250 Pack
1 options
Catalog No. 22544233 Supplier Biomedical Polymers Supplier No. BMP HEMODROP
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A simple, safe way to prepare differential slide preparations
  • No need to uncap specimen tubes
  • Smooth insertion into tube
  • Direct access to sample
  • Blood droplets visible on slide
  • Limits exposure to blood borne pathogens
Specifications
Product Type Blood Dispenser
Disposable Single-use
For Use With (Application) Prepare differential slide preparations safely and easily with direct access to the sample
Quantity 250 Pack