Learn More
Thermo Scientific™ BioMate™ 160 UV-Vis Spectrophotometer PROMO
Perform automated measurements of nucleic acids, proteins and cell cultures with the Thermo Scientific™ BioMate™ 160 UV-Vis spectrophotometer.
Supplier: Thermo Scientific™ 8403010002PM25
Description
The BioMate UV-Vis Spectrophotometer provides automation for high-throughput options and room-light resistance, allowing lid-open operation for improved speed and convenience. Software includes pre-programmed life science methods such as Nucleic Acid A260; Protein A280 and A205; Protein Colorimetric assays and OD600.
A full line of accessories, including automated cell changers and cell holders, sipper, and fiber optic probes are designed to simplify sampling and accommodate high throughput measurements and temperature control.
Features:
- 7-inch high-resolution touchscreen user interface
- Pre-programmed life science methods
- Networking capable (Wi-Fi adaptor available)
- Room light resistance for open-lid operation
- Sipper accessory
- Removable, washable sample compartment liner with magnetic placement and hold-down
- Accommodates cells up to 100 mm pathlength
- (Optional) 8-position cell changer with switchable turret
- (Optional) 4-position cell changer (long path cells)
- (Optional) Peltier-thermostatted single cell holder (20°C to 60°C)
- (Optional) Accessory for test-tubes up to 25 mm diameter and 150 mm height
Designed for:
- Advanced teaching labs
- R and D departments
- Applications requiring high throughput or temperature control
Specifications
| ±0.002 A | |
| Single USB-A; Duplex USB-A; Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available; Print via USB, Ethernet or Wi-Fi USB adaptor available | |
| 7 in. High Definition Color Touchscreen, Tiltable, 800 x 1280 pixels | |
| 2 nm | |
| 190 to 1100 nm | |
| External AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100/240 V, 50/60 Hz | |
| > 5 years typical, 3 years guaranteed | |
| Dual Beam | |
| -3 A to +5 A | |
| ±0.001 A at 1 A measured at 1.0 A at 546 nm | |
| Slow, medium and fast - up to 1600 nm/min. | |
| 2 Years + 1 Year extension with registration | |
| 0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm | |
| 7.5 kg |
| Accommodates (optional): 8-position cell changer, 4-position cell changer (long path cells, Peltier thermostatted cell holder (20 to 60°C), Sipper accessory, Fiber optic probe coupler | |
| Dual Silicon Photodiodes | |
| 35.5 x 38.5 x 19.5 cm | |
| UV-Vis Spectrophotometer | |
| <0.0005 A/hr., at 500 nm after 1 hour warmup | |
| Touchscreen | |
| ≤0.00020A at 0A at 260 and 500 nm, ≤0.00030A at 1A at 260 and 500 nm, ≤0.00040A at 2A at 260 and 500 nm | |
| ±0.002 A at 0.5 A, ±0.004 A at 1.0 A, ±0.008 A at 2.0 A | |
| -2 A to +3.5 A | |
| Snap-on printer available | |
| <1.0%T 198 nm (KCl), <0.05%T at 220 nm (NaI), <0.03%T at 340 nm (NaNO2) | |
| ±0.5 nm | |
| < ±0.2 nm |