missing translation for 'onlineSavingsMsg'
Learn More

bioMérieux™ VITEK™ Test Cards

Catalog No. B21343
Encompass
Change view
Click to view available options
For Use With (Application):
Gram positive cocci
Yeast
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
B21343 Yeast
B21342 Gram positive cocci
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21343 Supplier bioMérieux™ Supplier No. 21343

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Species-level identification and resistance detection in bacteria and yeast

  • Inoculation with a simple, standardized saline suspension of organism
  • No reagent addition
  • Closed and disposable for optimal user safety
  • Pre-applied bar codes for maximum traceability
  • Highest level of automation

Specifications

Product Type Identification Test
For Use With (Application) Yeast
For Use With (Equipment) Vitek™ 2 colorimetric
Quantity 20 Pack