missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Biohazard Labels Vinyl BIOHAZARD
Description
- Good resistance to most common chemicals, oils and water
- Warning labels support compliance with OSHA 29 CFR 1910.1030 Regulation for Bloodborne Pathogens Labeling
- Warning labels support compliance with DOT 49 CFR 172.400(b) Labeling Requirements for transportation
- Service temperature of -40°F to +122°F (-40°C to +50°C)
Specifications
Specifications
| Adhesive Type | Acrylic |
| Color | Black on Orange |
| Finish | Gloss |
| Format | Label |
| Material | Vinyl |
| Product Type | Label |
| Sterility | Non-sterile |
| No. per Pack | 25 Pk. |
| Dimensions (L x W) | 2.875 x 2 in. |
| Width (English) | 2 in. |