missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific™ Biohazard Disposal Bags

Description
- Biohazard collection bag
- Biohazard warning symbol and precautions printed in English, Spanish, and French
- Includes an indicator patch that darkens when autoclaved
- Autoclavable to 250°F (121°C)
- Disposable
- HS10044H bag holder is ONLY for use with HS10320 2 L bag
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | Red |
| Disposable | Single-use |
| For Use With (Application) | Biohazard Disposal |
| For Use With (Equipment) | Autoclave |
| Material | Polypropylene |
| Sterility | Non-sterile |
| No. per Case | 500 Pk. |
| Outer Packaging | Box |
| Thickness (English) | 63 mil |
| Show More |