missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poltex™ Biohazard Bag Box
Description
- Comes in two sizes to fit either 8 x 12 in. or 14 x 19 in. biohazard bags
- Constructed from super durable and cleanable PETG
- Rubber bumpons on base for non-slip surface
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Color | Clear |
| Holds | Bags |
| Material | PETG |
| Positions | 1 |
| Shape | Rectangular |
| Stackable | No |
| Sterility | Non-sterile |
| Length (Metric) | 102 mm |
| Product Type | Bag Rack |
| Show More |