missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ 96-Well, Gelatin-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Color: Clear; Sterility: Sterile; Lid: With Lid

Supplier:  Corning™ 354689

 View more versions of this product

Catalog No. CB354689


Only null left

Description

Description

Corning BioCoat Gelatin cultureware provides an attachment and growth promoting substrate for the culture of a variety of cell types. Gelatin is commonly used in the culture of vascular endothelial cells, muscle, embryonic stem (ES) cells, and F9 teratocarcinoma cells. It is also suitable for promoting adhesion of transfected cell types. Gelatin is a heterogeneous mixture of water-soluble proteins derived through the hydrolysis of Collagen.

Applications include:

  • Promotion of cell attachment and spreading
  • Culture of normal and transfected F9 teratocarcinoma cells for gene expression studies
  • Culture of HUVEC for E-Selectin7 expression and VEGF induction8

Source

  • Gelatin, porcine
  • Quality Control

    • Tested for ability to promote proliferation of HUVECs
    • Tested and found negative for bacteria and fungi

    Storage

  • 4°C to 30°C under dry conditions.
  • Order Info

    Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates

    Specifications

    Chemical Identifiers

    Standard
    4°C to 30°C
    8 x 12
    Polystyrene
    Sterile
    Round, Flat Bottom (F-well)
    Gelatin
    With Lid
    0.32 cm2
    Clear
    96
    Gelatin
    Promote cell attachment, spreading, proliferation and differentiation

    Specifications

    Standard
    4°C to 30°C
    8 x 12
    Clear
    96
    Gelatin
    Promote cell attachment, spreading, proliferation and differentiation
    Gelatin
    With Lid
    0.32 cm2
    5/Cs
    Polystyrene
    Sterile
    Round, Flat Bottom (F-well)
    5/Cs.
    Product Suggestions

    Product Suggestions

    Videos
    Safety and Handling

    Safety and Handling

    ShelfLife : 6 Months

    SDS
    Documents

    Documents

    Promotions

    Promotions