missing translation for 'onlineSavingsMsg'
Learn More

Ansell BioClean-D™ BDLC Sterile Lab Coats

Catalog No. p-7229237
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17010333 Small
17010332 Medium
17010331 Large
17010334 X-Large
17010335 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17010333 Supplier Ansell Supplier No. SBDLCS
Only null left

Manufactured from antistatic lightweight CleanTough material and features press stud fastenings, open cuffs, three pockets, and a rear vent.

  • Lightweight CleanTough material
  • Press stud fastening
  • Open cuffs and rear vent
  • Three deep pockets

Specifications

Product Type General Purpose Lab Coat
Size Small
Gender Unisex
Material Polyethylene, Non-Woven Polypropylene Laminate
Color White
Closure Type Snaps
Cuff Style Open
Disposable Limited-use
Sterility Sterile
Resistance Abrasion, Tears, Liquids, Chemicals
Style Knee-length
No. of Pockets 3
Latex Free Yes
Model S-BDLC
Seam Type Bound
Certifications/Compliance ASTM F739, ISO 11137-1
Certifications/Compliance (Europe) EN 1149-5, EN 13934-1, EN 13935-2, EN 530, EN 6530, EN 7854, EN 863, EN 9073-4, EN ISO 14325
For Use With (Application) Laboratory, Research, Veterninary Services
Autoclavable Not Autoclavable
Construction Bound Seams, Single Needle Stitching
Show More Show Less