missing translation for 'onlineSavingsMsg'
Learn More

Ansell BioClean-C™ Protective Aprons with Sleeves BCAS

Catalog No. p-7162362
Change view
Click to view available options
Size:
Small
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19668248 Small
19668249 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Please see product validation pack or contact Ansell customer service for specific data on use of garments with cytotoxic drugs. Garments used for protection against such drugs must be selected specifically for the type of chemicals used.
Catalog No. 19668248 Supplier Ansell Supplier No. BCASS
Only null left
Please see product validation pack or contact Ansell customer service for specific data on use of garments with cytotoxic drugs. Garments used for protection against such drugs must be selected specifically for the type of chemicals used.

Non-sterile apron with protective sleeves is an ideal choice when administering drugs, handling chemicals, and blending or compounding solids and liquids. Tested against a wide range of chemotherapy drugs.

  • Tested against permeation standard
  • ASTM F739-12
  • Ultrasonically bonded seams with protective tape
  • Tie tapes at rear
  • Adjustable neck fastening
  • Lightweight low-linting CleanTough material
  • Silicone-free
  • Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications
  • Conforms to Category 3 - Personal Protective Equipment Regulation (EU) 2016/425
  • Tested against standard EN ISO 13034:2005 + A1:2009 protective clothing against liquid chemicals (Type PB [6])
  • Tested against standard EN 14325 protective clothing against chemicals
  • Tested against permeation standard ASTM F739-12
  • Processed in an NEBB certified ISO Class 4 environment
  • Tested against standards: EN 530, Abrasion resistance; EN 6530, Resistance to penetration by liquids; EN 13934-1, Tensile strength; EN 9073-4, Trapezoidal tear resistance; EN 863, Puncture resistance; and EN 13935-2, Seam strength

Specifications

Size Small
Material Polypropylene, Polyethylene
Color Blue
Disposable Single-use
Closure Type Waist Ties
Sterility Non-sterile
Cuff Style Elastic
Resistance Liquids
For Use With (Application) Critical and Controlled Environments
Certifications/Compliance ASTM F739
Seam Type Bound
Certifications/Compliance (Europe) CE 0598, EN 13934-1, EN 13935-2, EN 530, EN 6530, EN 7854, EN 9073-4, EN ISO13688:2013, EN 14605:2005 + A1:2009, EN ISO 14325, Category III, EN 13034:2005 + A1:2009
Show More Show Less

Five (5) years from date of manufacture. Store in a dry, cool place (≤40°C) away from direct sunlight and fluorescent light.