Learn More
Biochrom Bio32+ Sodium Oxidized Protein Hydrolysate Systems
Designed for the analysis of complex oxidised hydrolysates such as those obtained from food and feedstuffs. Animal feedstuff, pet food, and the human food industry.
Supplier: Biochrom 80600074
Includes: Oxidised Hydrolysate system or ccelerated Sodium Buffer system comprising instrument, analytical column with top up resin, prewash column, BioSYS operating software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit.
Requires :
- Oxygen free Nitrogen gas (99.99%) or Argon gas regulated to 5 bar
- Drainage facility
- 240V/100V, 50Hz/60Hz, 300VA mains supply
Description
System uses a sodium citrate-based buffer system. These samples contain up to 20 amino acids normally found in proteins. A buffer system using 3 sodium citrate buffers is sufficient for these samples.
Accurate: Superb separation, accurately identifies and quantifies free amino acids, as well as the amino acid composition of proteins and peptides.
Reliable: Minimal matrix interference and near baseline separation helps to reduce ambiguities in interpretation and increase confidence in diagnosis of inborn errors of metabolism. Automated derivatization of amino acids maximizes reproducibility.
Fast: Rapid protocols that increase sample throughput by cutting analysis time.
Economical: The column cleaning and repacking service, gives columns an almost indefinite life and cuts the cost of ownership.
Fully Automated: Total PC control allows 24/7 operations.
Versatile: Wide variety of sample types compatible: Urine, plasma, serum, CSF, amniotic fluid, culture media, tissue extracts, protein hydrolysates from various food and feedstuff samples or synthetic drugs, nutraceuticals, beverages.
Easy to implement: No lengthy method development required as this well characterized system is established worldwide. All customers have their instrument optimised to their application directly from the factory.
Convenient: Interchangeable sample trays allow samples to be prepared in advance and stored refrigerated.
Robust: Advanced fluidics and robust hardware mean easy maintenance for users, backed by the reassurance of Biochrom's dedicated service and support team.
Flexible: Integrated software, choice of different methods to adapt to the user's needs. Automatic sample set-up. Networking and exporting of data.
Compact: Bench top design requiring minimum lab space.
Automatic shut-down and reaction coil flushing in the event of:
- Power failure
- Photometer lamp failure
- Low ninhydrin, buffer, coil and nitrogen pressures
- High coil and column temperatures, high ninhydrin, buffer and coil pressures
Full screening methods are available
361030934
Oxidised Hydrolysate system comprising instrument with 84 position cooled autosampler, Analytical column with top up resin, prewash column, BioSYS operating software, Ezchrom Elite data manipulation software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit. System also includes Pentium PC, 19 in. flat screen monitor and Deskjet printer
361030935
Oxidised Protein Hydrolysate instrument comprising 84-position cooled autosampler, analytical column with top up resin, prewash column, BioSYS operating software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit.
361030936
Bio 30+ main unit with analytical column with top up resin, prewash column, BioSYS operating software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit. Does not include Autosampler
361030937
Oxidised Protein Hydrolysate instrument comprising 84-position non-cooled autosampler, analytical column with top up resin, prewash column, BioSYS operating software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit.
361030938
Oxidised Protein Hydrolysate instrument comprisingh 84-position cooled autosampler, analytical column with top up resin, prewash column, BioSYS operating software, Ezchrom Elite data manipulation software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit. System also includes Pentium PC, 19 in. flat screen monitor and Deskjet printer. (High Resolution)
361030939
Oxidised Protein Hydrolysate instrument comprisingh 84-position cooled autosampler, analytical column with top up resin, prewash column, BioSYS operating software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit. (High Resolution)
361030940
Oxidised Protein Hydrolysate instrument comprising 84-position non-cooled autosampler, analytical column with top up resin, prewash column, BioSYS operating software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit. (High Resolution)
361030941
Accelerated Sodium Buffer System comprising instrument, 84-position cooled autosampler, analytical column with top up resin, prewash column, BioSYS operating software, Ezchrom Elite data manipulation software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit. System also includes Pentium PC, 19 in. flat screen monitor and Deskjet printer.
361030942
Accelerated Sodium Buffer System comprising instrument, 84-position cooled autosampler, analytical column with top up resin, prewash column, BioSYS operating software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit.
361030943
Accelerated Sodium Buffer instrument, analytical column with top up resin, prewash column, BioSYS operating software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit. Does not include Autosampler
361030944
Accelerated Sodium Buffer instrument comprising 84-position non-cooled autosampler, analytical column with top up resin, prewash column, BioSYS operating software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit.
- Indication of origin and type of protein source
- Quality of meat
- Indication of spoilage e.g. analysis of amine in fish products
- Indication of microbial contamination
- Research of optimum diet formulation and nutritional value
- Identification of unwanted supplements
- Proof of quality and nutritional value of final product
- Nutritional labelling
- Monitoring product throughout the production process
Chemical Identifiers
| Buffer pressure: maximum 150 bar; Ninhydrin pressure: maximum 24 bar | |
| High pressure PEEK column packed with Ultropac 8 cation exchange resin. Peltier heating/cooling system | |
| Bench top Fluidics Cabinet: 110 lb.; Autosampler: 46 lb. |
| Ninhydrin - post column derivatization | |
| Column temperature variable between 20°C to 99°C. Reaction coil temperature adjustable between 40°C to 145°C (135°C is optimum) | |
| Bench top Fluidics Cabinet: 50 kg; Autosampler: 21 kg |
Specifications
| Buffer pressure: maximum 150 bar; Ninhydrin pressure: maximum 24 bar | |
| With 84-position cooled autosampler, Ezchrom Elite software, Pentium PC, 19 in. flat screen monitor and printer; High Resolution | |
| Column temperature variable between 20°C to 99°C. Reaction coil temperature adjustable between 40°C to 145°C (135°C is optimum) | |
| Bench top Fluidics Cabinet: 50 kg; Autosampler: 21 kg | |
| High Resolution |
| Ninhydrin - post column derivatization | |
| High pressure PEEK column packed with Ultropac 8 cation exchange resin. Peltier heating/cooling system | |
| Bench top Fluidics Cabinet: 110 lb.; Autosampler: 46 lb. | |
| Cooled Autosampler, Software, PC, Monitor, Printer |