missing translation for 'onlineSavingsMsg'
Learn More

Biochrom Bio30+ Lithium Physiological Systems

Dedicated chromatographic bench top system for Amino Acid Analysis

Supplier:  Biochrom 80600056

 View more versions of this product

Catalog No. 361030923

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Includes: Bio 30+ main unit, analytical column with top up resin, prewash column, BioSYS operating software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit

Requires :
  • Oxygen free Nitrogen gas (99.99%) or Argon gas regulated to 5 bar
  • Drainage facility
  • 240V/100V, 50Hz/60Hz, 300VA mains supply


Description

Description

System uses a combination of 5 lithium citrate buffers to achieve a full amino acid profile in less than 90 minutes using the new accelerated method

Accurate: Superb separation, accurately identifies and quantifies free amino acids, as well as the amino acid composition of proteins and peptides.
Reliable: Minimal matrix interference and near baseline separation helps to reduce ambiguities in interpretation and increase confidence in diagnosis of inborn errors of metabolism. Automated derivatization of amino acids maximizes reproducibility.
Fast: Rapid protocols that increase sample throughput by cutting analysis time.
Economical: The column cleaning and repacking service, gives columns an almost indefinite life and cuts the cost of ownership.
Fully Automated: Total PC control allows 24/7 operations.
Versatile: Wide variety of sample types compatible: Urine, plasma, serum, CSF, amniotic fluid, culture media, tissue extracts, protein hydrolysates from various food and feedstuff samples or synthetic drugs, nutraceuticals, beverages.
Easy to implement: No lengthy method development required as this well characterized system is established worldwide. All customers have their instrument optimised to their application directly from the factory.
Convenient: Interchangeable sample trays allow samples to be prepared in advance and stored refrigerated.
Robust: Advanced fluidics and robust hardware mean easy maintenance for users, backed by the reassurance of Biochrom's dedicated service and support team.
Flexible: Integrated software, choice of different methods to adapt to the user's needs. Automatic sample set-up. Networking and exporting of data.
Compact: Bench top design requiring minimum lab space.

Automatic shut-down and reaction coil flushing in the event of:

  • Power failure
  • Photometer lamp failure
  • Low ninhydrin, buffer, coil and nitrogen pressures
  • High coil and column temperatures, high ninhydrin, buffer and coil pressures

Full screening methods are available
361030917
Bio 30+ main unit with 84-position cooled autosampler, analytical column with top up resin, prewash column, BioSYS operating software, Ezchrom Elite data manipulation software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit. System also includes Pentium PC, 19 in. flat screen monitor and Deskjet printer. (Accelerated Program)

361030918
84-position cooled autosampler, analytical column with top up resin, prewash column, BioSYS operating software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit. (Accelerated Program)

361030919
Bio 30+ main unit with analytical column with top up resin, prewash column, BioSYS operating software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit. Does not include Autosampler (Accelerated Program)

361030920
Bio 30+ main unit with 84-position cooled autosampler, analytical column with top up resin, prewash column, BioSYS operating software, Ezchrom Elite data manipulation software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit. System also includes Pentium PC, 19 in. flat screen monitor and Deskjet printer. (High Performance Program)

361030921
Bio 30+ main unit with 84-position cooled autosampler, Analytical column with top up resin, prewash column, BioSYS operating software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit. (High Performance Program)

361030922
Bio 30+ main unit with analytical column with top up resin, prewash column, BioSYS operating software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit. Does not include Autosampler (High Performance)

361030923
Bio 30+ main unit with 84-position cooled autosampler, analytical column with top up resin, prewash column, BioSYS operating software, Ezchrom Elite data manipulation software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit. System also includes Pentium PC, 19 in. flat screen monitor and Deskjet printer. (High Resolution Program)

361030924
Bio 30+ main unit with 84-position cooled autosampler, analytical column with top up resin, prewash column, BioSYS operating software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit. (High Resolution Program)

361030925
Bio 30+ main unit with analytical column with top up resin, prewash column, BioSYS operating software, chemical kit with calibration standard, Ninhydrin & buffer bottles, instruction manuals, tool kit and spare parts/consumables kit. Does not include Autosampler (High Resolution)

  • Detection and monitoring of metabolic disease and inborn errors of metabolism (IEM), such as PKU and MSUD and other aminoacidopathies in urine, plasma and CSF
  • Protein and peptide analysis for pharmaceutical applications
  • Food, beverages, plant extracts, fermentation products, culture media, nutraceuticals and infusion fluids

Specifications

Chemical Identifiers

Buffer pressure: maximum 150 bar; Ninhydrin pressure: maximum 24 bar
Ninhydrin - post column derivatization
Column temperature variable between 20°C to 99°C. Reaction coil temperature adjustable between 40°C to 145°C (135°C is optimum)
Bench top Fluidics Cabinet: 50 kg; Autosampler: 21 kg
IC System
High pressure PEEK column packed with Ultropac 8 cation exchange resin. Peltier heating/cooling system
Bench top Fluidics Cabinet: 110 lb.; Autosampler: 46 lb.

Specifications

Buffer pressure: maximum 150 bar; Ninhydrin pressure: maximum 24 bar
Ninhydrin - post column derivatization
High pressure PEEK column packed with Ultropac 8 cation exchange resin. Peltier heating/cooling system
Bench top Fluidics Cabinet: 110 lb.; Autosampler: 46 lb.
Autosampler, PC, Monitor, Printer
IC System
84-position cooled autosampler, Pentium PC, 19 in. flat screen monitor & Deskjet printer; High Resolution Program
Column temperature variable between 20°C to 99°C. Reaction coil temperature adjustable between 40°C to 145°C (135°C is optimum)
Bench top Fluidics Cabinet: 50 kg; Autosampler: 21 kg
High Resolution Program
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions