missing translation for 'onlineSavingsMsg'
Learn More

BioAcryl-P (40%, 39:1)

Catalog No. AAJ63323K2
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63323K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63323K2 Supplier Thermo Scientific Chemicals Supplier No. J63323K2

Acrylamide is used for sequencing gels and separation of proteins, used in protein and nucleic acid electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acrylamide is used for sequencing gels and separation of proteins, used in protein and nucleic acid electrophoresis.

Solubility
Fully miscible in water.

Notes
Protect from heat. Store away from oxidizing agents.

Specifications

Chemical Name or Material Acrylamide/Bisacrylamide 39:1
Physical Form Liquid
Concentration 40% soln
Color Colorless
Quantity 1 L
UN Number UN3426
Solubility Information Fully miscible in water.

RUO – Research Use Only