missing translation for 'onlineSavingsMsg'
Learn More

BioAcryl-P (30%, 37.5:1)

Catalog No. p-7042153
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61505AP 500 mL
AAJ61505K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61505AP Supplier Thermo Scientific Chemicals Supplier No. J61505AP
Only null left

BioAcryl-P (Acrylamide/Bisacrylamide 37.5:1, CAS # 25034-58-6) is a powder-based formula used to create the gel matrix necessary for electrophoresis experiments.

Acrylamide/Bisacrylamide 37.5:1, Premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • BioAcryl-P is a 37.5:1 ratio of Acrylamide/Bisacrylamide
  • Acrylamide is a crystalline amide that forms polymer structures at room temperature
  • Acrylamide acts as the monomer for the gel structure, and bisacrylamide as the crosslinker for gel matrix formation

Application

  • Suitable for gel electrophoresis-mediated separation of proteins and DNA for identification or sequencing

Specifications

Chemical Name or Material Acrylamide/Bisacrylamide 37.5:1
Physical Form Liquid
Concentration 30% soln
Color Colorless
Quantity 500 mL
MDL Number MFCD00080848
UN Number UN3426
Solubility Information It is soluble in water.

RUO – Research Use Only