missing translation for 'onlineSavingsMsg'
Learn More

BioAcryl-P (30%, 29:1)

BioAcryl-P (Acrylamide/Bisacrylamide 29:1, CAS # 25034-58-6) is a powder-based formula used to create the gel matrix necessary for electrophoresis experiments.

$300.10 - $485.47

Specifications

Chemical Name or Material Acrylamide/Bisacrylamide 29:1
Physical Form Liquid
Concentration 30% soln
Color Colorless
MDL Number MFCD00080848
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63279AP
View Documents
Thermo Scientific Chemicals
J63279AP
500 mL
Each for $300.10
Only null left
 
AAJ63279K2
View Documents
Thermo Scientific Chemicals
J63279K2
1 L
Each for $485.47
Only null left
 
Description

Description

Acrylamide is used for sequencing gels and separation of proteins, used in protein and nucleic acid electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• BioAcryl-P is a 29:1 ratio of Acrylamide/Bisacrylamide
• Acrylamide is a crystalline amide that forms polymer structures at room temperature
• Acrylamide acts as the monomer for the gel structure, and bisacrylamide as the crosslinker for gel matrix formation

Application

• Suitable for gel electrophoresis-mediated separation of proteins and DNA for identification or sequencing

Specifications

Specifications

Acrylamide/Bisacrylamide 29:1
30% soln
MFCD00080848
Difficult to mix.
Liquid
Colorless
UN3426
Videos
SDS
Documents

Documents

RUO – Research Use Only