missing translation for 'onlineSavingsMsg'
Learn More

BioAcryl-P (30%, 29:1)

Catalog No. AAJ63279AP
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63279AP 500 mL
AAJ63279K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63279AP Supplier Thermo Scientific Chemicals Supplier No. J63279AP

BioAcryl-P (Acrylamide/Bisacrylamide 29:1, CAS # 25034-58-6) is a powder-based formula used to create the gel matrix necessary for electrophoresis experiments.

Acrylamide is used for sequencing gels and separation of proteins, used in protein and nucleic acid electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• BioAcryl-P is a 29:1 ratio of Acrylamide/Bisacrylamide
• Acrylamide is a crystalline amide that forms polymer structures at room temperature
• Acrylamide acts as the monomer for the gel structure, and bisacrylamide as the crosslinker for gel matrix formation

Application

• Suitable for gel electrophoresis-mediated separation of proteins and DNA for identification or sequencing

Specifications

Chemical Name or Material Acrylamide/Bisacrylamide 29:1
Physical Form Liquid
Concentration 30% soln
Color Colorless
Quantity 500 mL
MDL Number MFCD00080848
UN Number UN3426
Solubility Information Difficult to mix.

RUO – Research Use Only