missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ Timothy Cubes, Certified DFS Item

Excellent source of fiber

Supplier:  Bio-Serv™ F1011

Catalog No. 14726357


Only null left
Explore available promotions

Description

Description

  • 100% sun-cured timothy hay
  • Steam pressed (no binders added)
  • Nutritionally assayed
  • Contaminant screened
  • Size: 3.18 x 1.90cm (1.25 x 0.75 in.)
  • 8.63kg (19 lb.)/Box
Specifications

Specifications

Timothy Cubes
100% Timothy Hay
Cubes
Cool dry place
1.91
Cubes
8630 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions