missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ Rodent BLT's™ Diet

Catalog No. 14726613
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14726613 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14726613 Supplier Bio-Serv™ Supplier No. F05072

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Feed as an appetite stimulant, treat or sole-source diet

  • Nutritionally complete, grain-based bacon-flavored diet
  • Precise 5g size eliminates guesswork for measuring food intake
  • Excellent for adding medications and test articles
  • Nutritionally assayed
  • 5g wafer, Dia. x H: 1 x 0.25 in. (2.5 x 0.6cm)
  • Can be customized to incorporate medication or test articles (contact your Fisher Scientific™ Customer Service Representative for details)
  • Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Animal Fat (Preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Magnesium Stearate, Calcium Silicate, Bacon Flavor, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin

Specifications

Product Type Rodent BLT's Diet
Components Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Animal Fat (Preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Magnesium Stearate, Calcium Silicate, Bacon Flavor, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin
Flavor Bacon
Particle Shape Wafer
Quantity 5 g
Species Rodent
Storage Requirements Cool dry place
Diameter (English) 1 in.
Diameter (Metric) 2.54 cm