missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bio-Serv™ Nutra-Gel Diet™ Purified Formula, Sterile
Description
- Ready to serve—no mixing required
- Provides both food and water
- Phytoestrogen free
- Bacon flavor stimulates appetite
- Each cell is individually sealed to maintain moisture and sterility
- Ideal for chewing and mobility impairments, recovering and debilitated rodents, weanlings, and transportation
- Nutritionally assayed
- Ingredients: Water, Sucrose, Casein, Corn Starch, Soybean Oil, Gelatin, Cellulose, Mineral Mix, Vitamin Mix, Bacon Flavor, L-Cystine, Xanthan Gum, Sorbic Acid, Choline Bitartrate, tBHQ
Specifications
Specifications
| Product Type | Animal Diet |
| Calories | 1.57 |
| Components | Water, Sucrose, Casein, Corn Starch, Soybean Oil, Gelatin, Cellulose, Mineral Mix, Vitamin Mix, Bacon Flavor, L-Cystine, Xanthan Gum, Sorbic Acid, Choline Bitartrate, tBHQ |
| Flavor | Bacon |
| For Use With (Application) | Use for chewing and mobility impairments, recovering and debilitated Rodents, weanlings and transportation |
| Particle Shape | Gel |
| Quantity | 16 |