missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bio-Serv™ Love Mash™ Rodent Reproductive Diet, Sterile
Description
- Feed as a supplement to regular diet
- Data shows increased litter size and pup survival rates
- Proven effective even in the most difficult strains
- Successfully used at many large breeding facilities
- Pellets are 0.5 in. dia. and vary in length
- Nutritionally assayed
- Ingredients: Oatmeal, Cod Liver Oil, Wheat Germ, Brewer's Yeast, Corn Syrup
Specifications
Specifications
| Product Type | Rodent Reproductive Diet |
| Calories | 3.83 |
| Components | Oatmeal, Cod Liver Oil, Wheat Germ, Brewer's Yeast, Corn syrup |
| Format | Pellet |
| Particle Shape | Pellet |
| Quantity | 1000 g |
| Species | Rodent |
| Storage Requirements | 6 Months, Refrigerated |
| Diameter (English) | 0.5 in. |
| Diameter (Metric) | 1.27 cm |