missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ Grain-Based Rodent Diet

Catalog No. 14726620
Encompass
Change view
Click to view available options
Quantity:
5000 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14726620 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14726620 Supplier Bio-Serv™ Supplier No. F3028

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used as a control diet when using custom grain-based diets

  • Nutritionally complete grain-based diet
  • Nutritionally assayed
  • Ingredients: Ground Corn, Dehulled Soybean Meal, Dried Beet Pulp, Fish Meal, Ground Oats, Brewers Dried Yeast, Cane Molasses, Dehydrated Alfalfa Meal, Dried Whey, Wheat Germ, Porcine Animal Fat (preserved with BHA), Porcine Meat Meal, Wheat Middlings, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride

Specifications

Product Type Control Diet
Calories 3.35
Components Ground Corn, Dehulled Soybean Meal, Dried Beet Pulp, Fish Meal, Ground Oats, Brewers Dried Yeast, Cane Molasses, Dehydrated Alfalfa Meal, Dried Whey, Wheat Germ, Porcine Animal Fat (preserved with BHA), Porcine Meat Meal, Wheat Middlings, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride
Format Pellet
For Use With (Application) Used as a control diet when ordering custom grain based diets
Particle Shape Pellet
Quantity 5000 g
Species Rodent
Storage Requirements Cool dry place
Diameter (English) 0.5 in.
Diameter (Metric) 1.27 cm
Show More Show Less