missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ Fruity Gems Mini Fruit-Flavored Cubes, Certified

Catalog No. 14726402
Encompass
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14726402 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14726402 Supplier Bio-Serv™ Supplier No. F5136

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A fruity treat for animals made with real papaya and pineapple

Use as training reward, treat or scatter in bedding to provide foraging activity.

  • Natural sweetness
  • Ingredients: papaya, pineapple, sugar, sulfur dioxide, citric acid, artificial flavors, food dye
  • Certified (contaminant screened)
  • 5kg/Box

Specifications

Product Type Animal Treats
Calories 3.52
Components Papaya, Pineapple, Sugar, Sulphur Dioxide, Citric Acid, Artificial Flavors, Food Dye
Flavor Fruit
Format Cubes
For Use With (Application) Use as training reward, treat or scatter in bedding to provide foraging activity
Particle Shape Cubes
Quantity Each
Storage Requirements Cool dry place