missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ Enrichment Treats, Mealworms, Certified

Catalog No. 14726508
Encompass
Change view
Click to view available options
Quantity:
16
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14726508 16
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14726508 Supplier Bio-Serv™ Supplier No. 9264

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Super premium freeze-dried meal worms

  • Healthy, natural source of fat and protein
  • Easy handling and delivery to animals
  • Approximately 1 in. long

Specifications

Product Type Mealworms
Particle Shape Mealworms
Quantity 16
Species Worms
Storage Requirements Cool dry place