missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ Enrichment Treats, Black Oil Sunflower Seeds, Sterile

Catalog No. 14726534
Encompass
Change view
Click to view available options
Quantity:
10,000 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14726534 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14726534 Supplier Bio-Serv™ Supplier No. S5137

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A healthy, natural foraging treat

An excellent source of omega 6 essential fatty acids, dietary fiber, protein, and vitamin E.

Due to the method used to process sunflower seeds, this package may contain other plant material.

Specifications

Product Type Sunflower Seeds Black Oil
Calories 4.48
For Use With (Application) Rodent
Particle Shape Oil
Quantity 10,000 g
Species Rodent
Storage Requirements Cool dry place