Learn More
Bio-Serv™ Crawl Balls™ Enrichment Device, Certified
Catalog No. 360993896
Encompass
Change view
Click to view available options
Color:
Amber
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
360993896 Amber
360993895 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 360993896 Supplier Bio-Serv™ Supplier No. K3330
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Provides a comfortable hiding place for rodents
  • 4 in. (10.2cm) dia. with three 2.5 in. (6.4cm) openings
  • Made of high-temperature polycarbonate
  • Transparent for ease of visibility
  • Autoclavable
  • Cagewasher safe
  • Certified (Contaminant screened)
Specifications
Autoclavable Autoclavable
Product Type Crawl Ball
Color Amber
Material Polycarbonate
Diameter (English) Inside 4 in.
Diameter (Metric) Inside 10.16 cm