missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ Bacon Softies™ Animal Diet, Certified

Catalog No. 14726703
Encompass
Change view
Click to view available options
Quantity:
1000 g
5000 g
Sterility:
Non-sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sterility
14726703 5000 g Non-sterile
14726701 1000 g Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14726703 Supplier Bio-Serv™ Supplier No. F3580

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Replaces labor intensive, nutrient diluted mush diets

  • Soft, pelleted, nutritionally rich diet for rodents and supplement for canines
  • Certified (contaminant screened)
  • Nutritionally assayed

Ingredients: Soy Flour, Soybean Oil, Corn Syrup, Sucrose, Casein, Cellulose, Corn Starch, Wheat Bran, Molasses, Mineral Mix, Banana Flakes, Acacia Gum, Bacon Flavor, Glycerin, Vitamin Mix, DL-Methionine, Choline Bitartrate, BHA

Specifications

Product Type Animal Diet
Components Soy Flour, Soybean Oil, Corn Syrup, Sucrose, Casein, Cellulose, Corn Starch, Wheat Bran, Molasses, Mineral Mix, Banana Flakes, Acacia Gum, Bacon Flavor, Glycerin, Vitamin Mix, DL-Methionine, Choline Bitartrate, BHA
Flavor Bacon
Format Pellet
For Use With (Application) For recovering or debilitated animals and weanlings
Particle Shape Soft Pellet
Quantity 5000 g
Species Rodent, Canine
Sterility Non-sterile
Diameter (English) 0.5 in.
Diameter (Metric) 1.27 cm
Show More Show Less