missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bio-Serv™ Bacon Softies™ Animal Diet, Certified
Replaces labor intensive, nutrient diluted mush diets
Supplier: Bio-Serv™ F35801
Description
- Soft, pelleted, nutritionally rich diet for rodents and supplement for canines
- Certified (contaminant screened)
- Nutritionally assayed
Ingredients: Soy Flour, Soybean Oil, Corn Syrup, Sucrose, Casein, Cellulose, Corn Starch, Wheat Bran, Molasses, Mineral Mix, Banana Flakes, Acacia Gum, Bacon Flavor, Glycerin, Vitamin Mix, DL-Methionine, Choline Bitartrate, BHA
Specifications
| Softies | |
| Bacon | |
| For recovering or debilitated animals and weanlings | |
| 1000 g | |
| Non-sterile | |
| 1.27 cm |
| Soy Flour, Soybean Oil, Corn Syrup, Sucrose, Casein, Cellulose, Corn Starch, Wheat Bran, Molasses, Mineral Mix, Banana Flakes, Acacia Gum, Bacon Flavor, Glycerin, Vitamin Mix, DL-Methionine, Choline Bitartrate, BHA | |
| Pellet | |
| Soft Pellet | |
| Rodent, Canine | |
| 0.5 in. |