missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ B-Wholesome Treats™ Animal Diet, Certified

Catalog No. 14726720
Encompass
Change view
Click to view available options
Quantity:
4500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14726720 4500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14726720 Supplier Bio-Serv™ Supplier No. F7500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Healthy alternative to the average biscuit

  • Whole wheat based
  • 100% USDA organic ingredients
  • Nutritionally assayed
  • Certified (contaminant screened)
  • 4.5kg/Box

Ingredients: Whole Wheat Flour, Extra Virgin Olive Oil, Dehydrated Whole Eggs, Non-Fat Dry Milk, Wheat Bran, Granulated Garlic, Honey

Specifications

Product Type Animal Treats
Calories 4.12
Components Whole Wheat Flour, Extra Virgin Olive Oil, Dehydrated Whole Eggs, Non-Fat Dry Milk, Wheat Bran, Granulated Garlic, Honey
Quantity 4500 g
Storage Requirements Cool dry place