missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Current Technologies Bio-Screen™ Biohazard Wipes, Liners, and Rolls
Low-linting, white, absorbent wipes bonded to an orange plastic backing
Supplier: Current Technologies BH16100
Description
- White absorbent side of this wipe / liner is low-linting and more absorbent than gauze or paper wipes
- Orange plastic backing of wipe / liner blocks soak through of blood, body fluids, specimens and chemicals (xylene, ketones, formalin) to gloves and surfaces
- Used for cleaning up small / large spills, to wipe pipets, probes, slide prep devices, and to pop caps from blood and specimen tubes
- Limit cross-contamination from gloves as employees touch phones, keyboards, doors, etc.
- Larger sizes can be used as mats and liners. Rolls of material can be cut to desired sizes
- Three thicknesses available: Standard (absorbs 9 times its weight), Heavy (absorbs 12 times its weight), Super (absorbs 25 times its weight)
Identifiants chimiques
| White and Orange | |
| Used for cleaning up small or large spills, to wipe pipets, probes, slide prep devices, and to pop caps from blood and specimen tubes. Wipes and Liners are non-sterile and not intended for use on patients. | |
| Each |
Spécifications
| Biohazard Wipe | |
| 100 ft. | |
| 16 in. | |
| 100 ft. x 16 in. | |
| Used for cleaning up small or large spills, to wipe pipets, probes, slide prep devices, and to pop caps from blood and specimen tubes. Wipes and Liners are non-sterile and not intended for use on patients. |
| Standard | |
| 30.4 m | |
| 406.4 mm | |
| White and Orange | |
| Each |
Wipes and Liners are non-sterile and not intended for use on patients.