Learn More
Bio-Foam™ Impression Kit
Catalog No. S99543
Change view
Click to view available options
High-density impression kit
Catalog No.
S99543 High-density impression kit
1 options
Catalog No. S99543 Supplier Customer Channels Group (Ccg) Supplier No. BIF100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Quick and easy-to-use system makes footwear impressions without damage or contamination.
  • Spray on a thin coating of Aerosol Dust & Dirt Hardener (sold separately)
  • Mix and pour in Shake-N-Cast™ ready-to-use casting material (sold separately, S99541)
  • Press shoe onto the surface
Specifications
For Use With (Application) To make permanent footwear impressions without damaging or contaminating the footwear
Includes Self-storing cardboard container
Product Type High Density Impression Kit