missing translation for 'onlineSavingsMsg'
Learn More

Dynalon™ Labware Bio-Bin™ Waste Disposal Containers

Catalog No. 17377646
Change view
Click to view available options
Capacity (Metric):
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
17377646 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17377646 Supplier Dynalon™ Labware Supplier No. 7973030005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The 5 L Bio-Bin™ is ideal for bench top use in labs as well as in wards and theaters. The bin comes folded flat which makes a significant space saving in storage and also makes the bin perfect for use in community nursing. All Bio-bin’s are fully autoclavable and are then disposed of according to their color or waste.

  • 96% paper based clinical and medical waste containers manufactured from FSC-certified sources.
  • Space efficient, light weight fold-flat design
  • Built-in lid and handles
  • Temporary and permanent seals
  • Stackable post use
  • UN 3291 approved
  • Printed user instructions and user ID/date information on lid
  • Waterproof construction
  • Built-in super absorbent Bio-matt™ to soak up any excess liquid in Bio-bin
  • Supplied with 2 x 2 in. Biohazard Waste Labels
  • Reorder information printed on the Bio-bin

Specifications

Capacity (Metric) 5 L
Capacity (English) 1.3 gal.
Color White
Dimensions (L x W x H) 7.2 x 7.2 x 14.6 in.
Quantity 50
Autoclavable Autoclavable
Material Cardboard
Product Type Disposal Container