missing translation for 'onlineSavingsMsg'
Learn More

Bindschedler's Green Leuco Base 98.0+%, TCI America™

Catalog No. p-7118473
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B04821G 1 g
B04825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B04821G Supplier TCI America Supplier No. B04821G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 637-31-0 | C16H21N3 | 255.365 g/mol

Chemical Identifiers

CAS 637-31-0
Molecular Formula C16H21N3
Molecular Weight (g/mol) 255.365
MDL Number MFCD00059291
InChI Key ZPGZNKGKCSYBGK-UHFFFAOYSA-N
PubChem CID 69473
IUPAC Name 1-N-[4-(dimethylamino)phenyl]-4-N,4-N-dimethylbenzene-1,4-diamine
SMILES CN(C)C1=CC=C(C=C1)NC2=CC=C(C=C2)N(C)C

Specifications

Melting Point 121°C
Synonym Bis(4-dimethylaminophenyl)amine, Leuco Bindschedler′Cs Green, N,N,N′C,N′C-Tetramethyldiaminodiphenylamine
Quantity 1 g
Physical Form Crystal/Powder
Formula Weight 255.37
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Bindschedler′s Green Leuco Base