missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bindschedler's Green Leuco Base 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 121°C |
| Synonym | Bis(4-dimethylaminophenyl)amine, Leuco Bindschedler′Cs Green, N,N,N′C,N′C-Tetramethyldiaminodiphenylamine |
| Quantity | 1 g |
| Physical Form | Crystal/Powder |
| Formula Weight | 255.37 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Chemical Name or Material | Bindschedler′s Green Leuco Base |
Safety and Handling
| TSCA | No |