missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Binding Buffer for Annexin V

Catalog No. 5018261
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5018261 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5018261 Supplier Invitrogen™ Supplier No. BMS500BB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Binding Buffer (4x) for Annexin V

50mL

Binding Buffer for Annexin V Apoptosis detection Kit

Reactivity/Species
Human, Mouse, Rat

Reported Application
Flow Cytometric Analysis

Specifications

Chemical Name or Material Binding Buffer
For Use With (Equipment) Flow Cytometer
Product Line eBioscience
Quantity 50 mL

For Research Use Only. Not for use in diagnostic procedures.