missing translation for 'onlineSavingsMsg'
Learn More

Bilobalide 95.0+%, TCI America™

Catalog No. B438810MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B438810MG 10 mg
B438850MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B438810MG Supplier TCI America Supplier No. B438810MG
Only null left

Chemical Identifiers

CAS 33570-04-6
Molecular Formula C15H18O8
Molecular Weight (g/mol) 326.30
MDL Number MFCD00132880,MFCD00238547
InChI Key MOLPUWBMSBJXER-YDGSQGCISA-N
PubChem CID 126963541
IUPAC Name (1S,4R,7R,8S,9R,11S)-9-tert-butyl-7,9-dihydroxy-3,5,12-trioxatetracyclo[6.6.0.0¹,¹¹.0⁴,⁸]tetradecane-2,6,13-trione
SMILES CC(C)(C)[C@]1(O)C[C@@H]2OC(=O)C[C@@]22C(=O)O[C@@H]3OC(=O)[C@H](O)[C@]123

Specifications

Color White
Quantity 10 mg
Formula Weight 326.30
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bilobalide
RTECSNumber LV1665000
TSCA No
Recommended Storage Freezer (-20 °C)