missing translation for 'onlineSavingsMsg'
Learn More

Fisher Science Education™ Bile Salts

Catalog No. S25196
Change view
Click to view available options
Quantity:
25 g
Packaging:
Solid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25196 25 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25196 Supplier Fisher Science Education™ Supplier No. S25196

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 83-44-3
Molecular Formula C24H40O4
Molecular Weight (g/mol) 392.58
MDL Number MFCD00003673
InChI Key KXGVEGMKQFWNSR-LLQZFEROSA-N
Synonym deoxycholic acid, deoxycholate, desoxycholic acid, choleic acid, cholerebic, cholorebic, degalol, deoxycholatic acid, droxolan, pyrochol
PubChem CID 222528
ChEBI CHEBI:28834
IUPAC Name (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
SMILES [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

Packaging Solid
Quantity 25 g
Concentration or Composition (by Analyte or Components) Sodium Salts of Clycoholic and Taurocholic Acids
Chemical Name or Material Bile Salts
Safety handling Check Alert storage code Red
ShelfLife 2 years