missing translation for 'onlineSavingsMsg'
Learn More

Bifonazole Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014864
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014864 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014864 Supplier Merck Emd Millipore Supplier No. PHR12961G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Bifonazole is a substituted imidazole analog with therapeutic potential in treating invasive mucosal infections. It exhibits antifungal activity by suppressing the proliferation of dermatophytes, yeasts and fungi affecting the skin and nails. Its mode of action involves blocking the fungal ergosterol biosynthetic pathway and additional inhibition of terpenoid biosynthesis. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 60628-96-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C22H18N2
MDL Number MFCD00865567
Synonym 1-(p,alpha-Diphenylbenzyl)imidazole
Type Certified Reference Material
Quantity 1 g
Formula Weight 310.39
Grade Certified Reference Material