missing translation for 'onlineSavingsMsg'
Learn More

Bifonazole, 98%, Thermo Scientific™

Catalog No. AC458620050
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458620050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC458620050 Supplier Thermo Scientific Chemicals Supplier No. 458620050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 60628-96-8
Loss on Drying 0.5% max. (1 g, 105°C, 1 h)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Molecular Formula C22H18N2
Quantity 5g
Synonym bifonazole, mycospor, trifonazole, bifonazol, bifonazolum, amycor, azolmen, bifonazol inn-spanish, bifonazolum inn-latin, 1-4-biphenylyl phenylmethyl-1h-imidazole
InChI Key OCAPBUJLXMYKEJ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(C3=CC=CC=C3)N4C=CN=C4
IUPAC Name 1-[phenyl-(4-phenylphenyl)methyl]imidazole
Molecular Weight (g/mol) 310.39
PubChem CID 2378
ChEBI CHEBI:78692
Formula Weight 310.39
Percent Purity 98%
Chemical Name or Material Bifonazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 262-336-6

RUO – Research Use Only