missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Bifenox

Catalog No. AC464600010
Catalog No. AC464600010 Supplier Thermo Scientific™ Supplier No. 464600010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 42576-02-3
Molecular Formula C14H9Cl2NO5
Molecular Weight (g/mol) 342.13
MDL Number MFCD00055314
InChI Key SUSRORUBZHMPCO-UHFFFAOYSA-N
IUPAC Name methyl 5-(2,4-dichlorophenoxy)-2-nitrobenzoate
SMILES COC(=O)C1=CC(OC2=CC=C(Cl)C=C2Cl)=CC=C1[N+]([O-])=O

Specifications

Solubility Information Solubility aq. soln.: insoluble.
Formula Weight 342.13
Chemical Name or Material Bifenox
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Very toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

EINECSNumber 255-894-7

RUO – Research Use Only