missing translation for 'onlineSavingsMsg'
Learn More

Biebrich Scarlet sodium salt

Catalog No. AAJ6560314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6560314 25 g
AAJ6560322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6560314 Supplier Thermo Scientific Chemicals Supplier No. J6560314
Only null left

CAS: 4196-99-0 | C22H14N4Na2O7S2 | 556.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4196-99-0
Molecular Formula C22H14N4Na2O7S2
Molecular Weight (g/mol) 556.47
MDL Number MFCD00003891
InChI Key YCKMEQRBEVVZQF-VKZXTWAVSA-L
Synonym C.I. 26905; Acid Red 66
PubChem CID 16219040
IUPAC Name disodium 2-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-[2-(4-sulfonatophenyl)diazen-1-yl]benzene-1-sulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C(=C1)S([O-])(=O)=O

Specifications

Melting Point 181°C to 188°C
Beilstein 3883900
Formula Weight 556.49
Color Brown to Red
Quantity 25 g
Flash Point 300°C (572°F)
Physical Form Powder
Chemical Name or Material Biebrich Scarlet sodium salt
EINECSNumber 224-084-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only