missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Biebrich Scarlet, dye content approx. 70%

Catalog No. AC402221000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC402221000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC402221000 Supplier Thermo Scientific Chemicals Supplier No. 402221000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4196-99-0 | C22H14N4Na2O7S2 | 556.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4196-99-0
Molecular Formula C22H14N4Na2O7S2
Molecular Weight (g/mol) 556.47
MDL Number MFCD00003891
InChI Key YCKMEQRBEVVZQF-VKZXTWAVSA-L
Synonym Acid Red 66, C.I. 26905, Ponceau BS, Croceine scarlet
PubChem CID 16219040
IUPAC Name disodium 2-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-[2-(4-sulfonatophenyl)diazen-1-yl]benzene-1-sulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C(=C1)S([O-])(=O)=O

Specifications

Melting Point 181.0°C to 188.0°C
Packaging Glass bottle
Assay Percent Range 99%
Solubility Information Solubility in water: soluble. Other solubilities: soluble in cellosolve, glycol, alcohol
Quantity 100 g
Formula Weight 556.48
Chemical Name or Material Biebrich Scarlet, 99%
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only